SY3H3D67D8FB Synthonix Corporation

Ethyl 3-chloro-4-formylbenzoate

  • MDL number MFCD21603681
  • Cas Number : 1465327-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54023.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54023.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart