SY3H3D67D8FC Synthonix Corporation

Ethyl 2-(4-chlorophenyl)-2-oxoacetate

  • MDL number MFCD01319615
  • Cas Number : 34966-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54025.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54025.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart