SY3H3D67D900 Synthonix Corporation

Ethyl imidazo[1,5-a]pyridine-7-carboxylate hydrochloride

  • MDL number MFCD29918421
  • Cas Number : 1914148-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54089.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54089.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart