SY3H3D67D902 Synthonix Corporation

Ethyl quinuclidine-4-carboxylate hydrochloride

  • Cas Number : 22766-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54207.html

Safety Information

MSDS https://synthonix.com/54207.html
Availability details for
Quantity
  • Availability
  • Add to Cart