SY3H3D67D904 Synthonix Corporation

ethyl 4-(aminomethyl)cyclohexane-1-carboxylate hydrochloride

  • MDL number MFCD28656551
  • Cas Number : 19878-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54214.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54214.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart