SY3H3D67D905 Synthonix Corporation

Ethyl 3-amino-3-(pyridin-3-yl)propanoate dihydrochloride

  • MDL number MFCD11053562
  • Cas Number : 149498-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54240.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54240.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart