SY3H3D67D907 Synthonix Corporation

Ethyl 3-bromo-1H-indole-2-carboxylate

  • MDL number MFCD02071793
  • Cas Number : 91348-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Slightly pale yellow - Greyish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54318.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54318.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart