SY3H3D67D908 Synthonix Corporation

Ethyl 5-bromo-1H-indole-7-carboxylate

  • MDL number MFCD21608002
  • Cas Number : 1065181-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54319.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54319.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart