SY3H3D67D95F Synthonix Corporation

Ethyl 6-bromo-4-oxo-3,4-dihydro-2H-benzo[e][1,3]oxazine-2-carboxylate

  • MDL number MFCD27952666
  • Cas Number : 30482-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54335.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54335.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart