SY3H3D67D965 Synthonix Corporation

Ethyl 3-(2,5-difluorophenyl)-3-oxopropanoate

  • MDL number MFCD07783808
  • Cas Number : 887267-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54360.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54360.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart