SY3H3D67D96B Synthonix Corporation

1-Ethyl-1H-indole-6-carbaldehyde

  • Cas Number : 854778-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54406.html

Safety Information

MSDS https://synthonix.com/54406.html
Availability details for
Quantity
  • Availability
  • Add to Cart