SY3H3D67D96F Synthonix Corporation

Ethyl 2-cyano-2-phenylacetate

  • MDL number MFCD00001866

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Very pale yellow - yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54414.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54414.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart