SY3H3D67D971 Synthonix Corporation

5-Ethoxy-1H-indole-3-carbaldehyde

  • MDL number MFCD07364715
  • Cas Number : 169789-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54417.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54417.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart