SY3H3D67D972 Synthonix Corporation

Ethyl 2-(benzo[d]thiazol-2-yl)acetate

  • MDL number MFCD00546397
  • Cas Number : 29182-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Pale yellow - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54427.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54427.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart