SY3H3D67D974 Synthonix Corporation

Ethyl 5-hydroxyindole-2-carboxylate

  • MDL number MFCD00015458
  • Cas Number : 24985-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to Yellow or Pale-red Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54432.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54432.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart