SY3H3D67D982 Synthonix Corporation

Ethyl 5-methylpyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number MFCD06739193
  • Cas Number : 51135-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54490.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54490.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart