SY3H3D67D984 Synthonix Corporation

Ethyl 4-amino-5-cyano-2-hydroxy-3-methylbenzoate

  • Cas Number : 72817-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54495.html

Safety Information

MSDS https://synthonix.com/54495.html
Availability details for
Quantity
  • Availability
  • Add to Cart