SY3H3D67D98F Synthonix Corporation

Ethyl 2,2-difluoro-2-(2-methoxyphenyl)acetate

  • Cas Number : 1150164-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54583.html

Safety Information

MSDS https://synthonix.com/54583.html
Availability details for
Quantity
  • Availability
  • Add to Cart