SY3H3D67D992 Synthonix Corporation

2-(2-Ethoxy-4-formylphenoxy)acetic acid

  • MDL number MFCD01835931
  • Cas Number : 51264-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54596.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54596.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart