SY3H3D67D9A2 Synthonix Corporation

Ethyl 2-methoxy-6-methylbenzoate

  • MDL number MFCD00191666
  • Cas Number : 6520-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Colorless - Pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54791.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54791.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart