SY3H3D67D9F6 Synthonix Corporation

Ethyl 2-(4-hydroxyphenoxy)propanoate

  • Cas Number : 65343-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54807.html

Safety Information

MSDS https://synthonix.com/54807.html
Availability details for
Quantity
  • Availability
  • Add to Cart