SY3H3D67D9F8 Synthonix Corporation

Ethyl 3-hydroxy-8,9-dihydro-5H-pyridazino[3,4-d]azepine-7(6H)-carboxylate

  • MDL number MFCD12756173
  • Cas Number : 1190897-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54845.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54845.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart