SY3H3D67DA12 Synthonix Corporation

Ethyl 3-(4-chlorophenyl)propiolate

  • MDL number MFCD00456510
  • Cas Number : 20026-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55405.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55405.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart