SY3H3D67DA13 Synthonix Corporation

Ethyl 7-chloro-3-hydroxybenzo[b]thiophene-2-carboxylate

  • Cas Number : 1825392-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55412.html

Safety Information

MSDS https://synthonix.com/55412.html
Availability details for
Quantity
  • Availability
  • Add to Cart