SY3H3D67DA26 Synthonix Corporation

Ethyl quinoline-4-carboxylate

  • Cas Number : 10447-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55661.html

Safety Information

MSDS https://synthonix.com/55661.html
Availability details for
Quantity
  • Availability
  • Add to Cart