SY3H3D67DA28 Synthonix Corporation

Ethyl isoquinoline-6-carboxylate

  • Cas Number : 188861-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55663.html

Safety Information

MSDS https://synthonix.com/55663.html
Availability details for
Quantity
  • Availability
  • Add to Cart