SY3H3D67DA35 Synthonix Corporation

2-(Ethoxycarbonyl)-1H-indole-3-carboxylic acid

  • MDL number MFCD18803524
  • Cas Number : 441800-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55687.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55687.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart