SY3H3D67DA3D Synthonix Corporation

Ethyl 1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD16584325
  • Cas Number : 115315-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Colorless Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55712.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55712.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart