SY3H3D67DA3E Synthonix Corporation

Ethyl 4-phenyl-1H-pyrazole-3-carboxylate

  • Cas Number : 6963-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55713.html

Safety Information

MSDS https://synthonix.com/55713.html
Availability details for
Quantity
  • Availability
  • Add to Cart