SY3H3D67DA45 Synthonix Corporation

Ethyl 5-amino-1-(2-chlorophenyl)-1H-pyrazole-4-carboxylate

  • Cas Number : 14678-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55735.html

Safety Information

MSDS https://synthonix.com/55735.html
Availability details for
Quantity
  • Availability
  • Add to Cart