SY3H3D67DA47 Synthonix Corporation

Ethyl 5-amino-1-(2-fluorophenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD03407403
  • Cas Number : 618070-65-8
  • Molecular Weight : 249.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-amino-1-(2-fluorophenyl)-1H-pyrazole-4-carboxylate
Inchi Code 1S/C12H12FN3O2/c1-2-18-12(17)8-7-15-16(11(8)14)10-6-4-3-5-9(10)13/h3-7H,2,14H2,1H3
Inchi Key NYHXOVWRDYMNGL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/55738.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/55738.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart