SY3H3D67DA4A Synthonix Corporation

Ethyl 5-amino-1-(4-nitrophenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00277862
  • Cas Number : 16459-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55743.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55743.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart