SY3H3D67DA4D Synthonix Corporation

Ethyl 5-methoxybenzo[b]thiophene-2-carboxylate

  • MDL number MFCD13180956
  • Cas Number : 40862-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55773.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55773.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart