SY3H3D67DA52 Synthonix Corporation

Ethyl 5-amino-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 866837-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55800.html

Safety Information

MSDS https://synthonix.com/55800.html
Availability details for
Quantity
  • Availability
  • Add to Cart