SY3H3D67DA59 Synthonix Corporation

Ethyl 5-methyl-3-(thiophen-2-yl)-1H-pyrrole-2-carboxylate

  • Cas Number : 31894-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55824.html

Safety Information

MSDS https://synthonix.com/55824.html
Availability details for
Quantity
  • Availability
  • Add to Cart