SY3H3D67DA5B Synthonix Corporation

Ethyl 1-(4-nitrophenyl)cyclopropanecarboxylate

  • MDL number MFCD22381468
  • Cas Number : 1308814-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55836.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55836.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart