SY3H3D67DA5D Synthonix Corporation

Ethyl 4-(2-nitrophenyl)-3-oxobutanoate

  • MDL number MFCD01926658
  • Cas Number : 66073-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55843.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55843.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart