SY3H3D67DA61 Synthonix Corporation

Ethyl 5-amino-4-cyano-3-(2-ethoxy-2-oxoethyl)thiophene-2-carboxylate

  • MDL number MFCD00832845
  • Cas Number : 58168-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Light-red or Pale yellow to Yellow Powder or Crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55880.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55880.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart