SY3H3D67DA68 Synthonix Corporation

Ethyl (4-Methoxybenzoyl)acetate

  • MDL number MFCD00449636
  • Cas Number : 2881-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55939.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/55939.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart