SY3H3D67DA69 Synthonix Corporation

Ethyl 3-(benzylamino)-3-oxopropanoate

  • MDL number MFCD28128094
  • Cas Number : 29689-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55991.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55991.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart