SY3H3D67DA76 Synthonix Corporation

Ethyl 2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate

  • MDL number MFCD10566154
  • Cas Number : 16323-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56172.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56172.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart