SY3H3D67DA7B Synthonix Corporation

(1-(6-Ethoxypyrimidin-4-yl)piperidin-3-yl)methanol

  • Cas Number : 1404192-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56232.html

Safety Information

MSDS https://synthonix.com/56232.html
Availability details for
Quantity
  • Availability
  • Add to Cart