SY3H3D67DA7E Synthonix Corporation

1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine

  • MDL number MFCD27665207
  • Cas Number : 253168-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56246.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56246.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart