SY3H3D67DA81 Synthonix Corporation

(S)-Ethyl-N-Boc-pyroglutamate

  • MDL number MFCD07778090
  • Cas Number : 144978-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56249.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56249.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart