SY3H3D67DA88 Synthonix Corporation

Ethyl 4-Boc-2-morpholinecarboxylate

  • MDL number MFCD01646390
  • Cas Number : 768371-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56284.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56284.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart