SY3H3D67DAD9 Synthonix Corporation

Ethyl 4-chloro-6,7-difluoroquinoline-3-carboxylate

  • MDL number MFCD06014061
  • Cas Number : 318685-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56448.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56448.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart