SY3H3D67DAE0 Synthonix Corporation

(4-(4-Ethylpiperazin-1-yl)phenyl)boronic acid monohydrochloride

  • Cas Number : 2096339-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56631.html

Safety Information

MSDS https://synthonix.com/56631.html
Availability details for
Quantity
  • Availability
  • Add to Cart