SY3H3D67DAEB Synthonix Corporation

4-Ethoxy-1-naphthoic acid

  • MDL number MFCD06260709
  • Cas Number : 19692-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56784.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56784.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart