SY3H3D67DAF2 Synthonix Corporation

Ethyl 3-(4-methoxyphenyl)isoxazole-5-carboxylate

  • Cas Number : 376623-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56830.html

Safety Information

MSDS https://synthonix.com/56830.html
Availability details for
Quantity
  • Availability
  • Add to Cart