SY3H3D67DAF7 Synthonix Corporation

Ethyl 1-methyl-3-phenyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD03933288
  • Cas Number : 10250-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56848.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56848.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart